Learn More
Cayman Chemical Metaproterenol (hemisulfate), Purity: ≥98%, CAS: 5874-97-5, 100 mg
SDP
Catalog No. 502994086
Encompass_Preferred
A long-acting β2-AR agonist with bronchodilator actions; inhibits TGF-α-induced hepatocyte DNA syntheses and proliferation in vitro (IC50 = 15 nM); dose-dependently relaxes lung strips and increases heart rate and inotropy of right atria and left atria, respectively, isolated from guinea pig (pD2s = 6.4, 7.08, and 6.61, respectively)
Catalog No. 50-299-4086 Supplier Cayman Chemical Supplier No. 23611100 MG
A long-acting β2-AR agonist with bronchodilator actions; inhibits TGF-α-induced hepatocyte DNA syntheses and proliferation in vitro (IC50 = 15 nM); dose-dependently relaxes lung strips and increases heart rate and inotropy of right atria and left atria, respectively, isolated from guinea pig (pD2s = 6.4, 7.08, and 6.61, respectively)