Learn More
Medchemexpress LLC Metaraminol tartrate | 33402-03-8 | 99.9% | 200 MG
SDP
Catalog No. 5000413070
Encompass_Preferred
Metaraminol tartrate is an α-adrenergic agonist and a sympathomimetic amine. It directly and indirectly affects adrenergic receptors, with a predominant alpha effect, and acts as a vasopressor agent.
  • Stimulation of α-adrenoceptors by metaraminol has been shown to accelerate cell proliferation in colonie crypts.
  • Lethal endotoxin shock was prevented in a study involving dogs.
  • It has been investigated in clinical trials for conditions such as preeclampsia, cesarean section complications, and hypotension.
Catalog No. 50-004-13070 Supplier Medchemexpress LLC Supplier No. HYB1116200MG
Metaraminol tartrate is an α-adrenergic agonist and a sympathomimetic amine. It directly and indirectly affects adrenergic receptors, with a predominant alpha effect, and acts as a vasopressor agent.
  • Stimulation of α-adrenoceptors by metaraminol has been shown to accelerate cell proliferation in colonie crypts.
  • Lethal endotoxin shock was prevented in a study involving dogs.
  • It has been investigated in clinical trials for conditions such as preeclampsia, cesarean section complications, and hypotension.