Learn More
Cayman Chemical MetaramInoltartrate 100mg
SDP
Catalog No. 502994386
Encompass_Preferred
A vasoconstrictor; increases cardiac output, mean arterial pressure, and coronary blood flow and decreases pulse rate in a canine model of cardiogenic shock when administered at a dose of 0.05 mg/kg; increases survival in a canine model of hemorrhagic hypotension; reverses acetoxycycloheximide-induced transient amnesia in a shock avoidance test in rats
Catalog No. 50-299-4386 Supplier Cayman Chemical Supplier No. 23938100 MG
A vasoconstrictor; increases cardiac output, mean arterial pressure, and coronary blood flow and decreases pulse rate in a canine model of cardiogenic shock when administered at a dose of 0.05 mg/kg; increases survival in a canine model of hemorrhagic hypotension; reverses acetoxycycloheximide-induced transient amnesia in a shock avoidance test in rats