Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metarrestin | 1443414-10-5 | 99.9% | 100 MG
SDP

Catalog No. 5000232036
Encompass_Preferred

Metarrestin is a first-in-class small-molecule inhibitor of the perinucleolar compartment (PNC) used in preclinical cancer research to disrupt nucleolar structure and inhibit RNA polymerase I transcription.

  • First-in-class perinucleolar compartment inhibitor for preclinical research.
  • Disrupts nucleolar structure and inhibits RNA polymerase I transcription.
  • Molecular formula C31H30N4O; molecular weight 474.60.
  • Purity 99.9% (reported by supplier).
  • Available as a 100 MG powder; store powder at -20°C (long term) or 4°C.
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-32036 Supplier Medchemexpress LLC Supplier No. HY120118100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metarrestin is a first-in-class small-molecule inhibitor of the perinucleolar compartment (PNC) used in preclinical cancer research to disrupt nucleolar structure and inhibit RNA polymerase I transcription.

  • First-in-class perinucleolar compartment inhibitor for preclinical research.
  • Disrupts nucleolar structure and inhibits RNA polymerase I transcription.
  • Molecular formula C31H30N4O; molecular weight 474.60.
  • Purity 99.9% (reported by supplier).
  • Available as a 100 MG powder; store powder at -20°C (long term) or 4°C.
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.