Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metarrestin | 1443414-10-5 | 99.9% | 50 MG
SDP

Catalog No. 5000232045
Encompass_Preferred

Metarrestin is a small-molecule perinucleolar compartment inhibitor provided as a high-purity research compound for in vitro and in vivo studies investigating nucleolar structure and metastasis.

  • Provides high purity (99.9%).
  • Molecular formula C31H30N4O and molecular weight 474.60 g/mol.
  • Available in solid and solution formats for flexibility in experiments.
  • Suitable for use in biochemical, cell-based, and pharmacokinetic studies.
  • Recommended storage: powder at -20°C; in solvent at -80°C.

Catalog No. 50-002-32045 Supplier Medchemexpress LLC Supplier No. HY12011850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metarrestin is a small-molecule perinucleolar compartment inhibitor provided as a high-purity research compound for in vitro and in vivo studies investigating nucleolar structure and metastasis.

  • Provides high purity (99.9%).
  • Molecular formula C31H30N4O and molecular weight 474.60 g/mol.
  • Available in solid and solution formats for flexibility in experiments.
  • Suitable for use in biochemical, cell-based, and pharmacokinetic studies.
  • Recommended storage: powder at -20°C; in solvent at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.