Learn More
Selleck Chemical LLC Metarrestin S3472-5mg
SDP
Supplier:  Selleck Chemical LLC S34725MG
Encompass_Preferred
Metarrestin is an orally active and specific perinucleolar compartment (PNC) inhibitor that disrupts the nucleolar structure and inhibits RNA polymerase (Pol) I transcription Metarrestin blocks metastatic development and extends survival in mouse cancer models
Catalog No. 50-217-3117