Learn More
Metasystems XCE Y Hybridization Probe
SDP
Catalog No. 22200368
Encompass
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
22-200-368 50 μL
1 options
Catalog No. 22-200-368 Supplier Metasystems Supplier No. D0824050BL
Specifications
For Use With (Application) Produce sharp and bright signals allowing for rapid and precise identification and enumeration of Human chromosomes in metaphase and interphase cells on various sample types
Quantity 50 μL