Learn More
Metasystems XL ATM/TP53 Hybridization Probe
SDP
Catalog No. 22200321
Encompass
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
22-200-321 100 μL
1 options
Catalog No. 22-200-321 Supplier Metasystems Supplier No. D5046100OG
Specifications
For Use With (Application) Detects deletions in the long arm of chromosome 11 and in the short arm of chromosome 17