Learn More
Metasystems XL DLEU/TP53 Hybridization Probe
SDP
Catalog No. 22200338
Encompass
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
22-200-338 100 μL
1 options
Catalog No. 22-200-338 Supplier Metasystems Supplier No. D5067100OG
Specifications
For Use With (Application) Detects deletions in the long arm of chromosome 13 and in the short arm of chromosome 17