Learn More
ILEX LIFE SCIENCES LLC Metatissue hPLMA Sponge scaffolds for 96 well plate 3D cell culture 1/EA
SDP
Catalog No. 5000580275
Encompass_Preferred
Metatissue hPLMA Sponge - Human Methacryloyl Platelet Lysates Scaffold for 96-well 3D Cell Culture (48 ct ) INTRODUCTION Metatissue hPLMA Sponge is a human based xeno-free bioactive platelet lysates-derived platform that provides physiologically relevant microenvironments for cell culture hPLMA Sponge is a porous scaffold ready to use with high reproducibility and increased area for cell growth and migration hPLMA Sponge is designed to be suitable for 96 well-plates and no photoinitiator or light source is needed hPLMA Sponge is a biocompatible platform that supports cell attachment proliferation and differentiation even without traditional animal derived supplementation hPLMA Sponge derived from hPL is composed of several native proteins including growth factors and cytokines (PDGF VEGF EPO EGF HGF) These bioactive proteins are known to be key components involved in cell adhesion and proliferation processes and are major regulators of angiogenesis
Catalog No. 50-005-80275 Supplier ILEX LIFE SCIENCES LLC Supplier No. PL02
Metatissue hPLMA Sponge - Human Methacryloyl Platelet Lysates Scaffold for 96-well 3D Cell Culture (48 ct ) INTRODUCTION Metatissue hPLMA Sponge is a human based xeno-free bioactive platelet lysates-derived platform that provides physiologically relevant microenvironments for cell culture hPLMA Sponge is a porous scaffold ready to use with high reproducibility and increased area for cell growth and migration hPLMA Sponge is designed to be suitable for 96 well-plates and no photoinitiator or light source is needed hPLMA Sponge is a biocompatible platform that supports cell attachment proliferation and differentiation even without traditional animal derived supplementation hPLMA Sponge derived from hPL is composed of several native proteins including growth factors and cytokines (PDGF VEGF EPO EGF HGF) These bioactive proteins are known to be key components involved in cell adhesion and proliferation processes and are major regulators of angiogenesis