Learn More
Medchemexpress LLC Metaxalone | 1665-48-1 | 99.8% | 50 MG
SDP
Catalog No. 5000408231
Encompass_Preferred
Metaxalone is an FDA-approved muscle relaxant that primarily acts on the central nervous system by inhibiting polysynaptic reflex arcs. It also functions as an inhibitor of MAO-A, demonstrating anti-inflammatory and antioxidant effects. This product is for research use only.
  • Inhibits polysynaptic reflex arcs
  • Acts as an inhibitor of MAO-A
  • Demonstrates anti-inflammatory and antioxidant effects
  • Inhibits IL-1β-induced inflammatory phenotype
  • Modulates NF-κB and other related signaling pathways
  • Decreases MAO-A expression and activity in IL-1β-treated microglia
Catalog No. 50-004-08231 Supplier Medchemexpress LLC Supplier No. HYB067850MG
Metaxalone is an FDA-approved muscle relaxant that primarily acts on the central nervous system by inhibiting polysynaptic reflex arcs. It also functions as an inhibitor of MAO-A, demonstrating anti-inflammatory and antioxidant effects. This product is for research use only.
  • Inhibits polysynaptic reflex arcs
  • Acts as an inhibitor of MAO-A
  • Demonstrates anti-inflammatory and antioxidant effects
  • Inhibits IL-1β-induced inflammatory phenotype
  • Modulates NF-κB and other related signaling pathways
  • Decreases MAO-A expression and activity in IL-1β-treated microglia