Learn More
Medchemexpress LLC Metaxalone-d3 | 1192812-66-0 | 98.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0678S1MG
Encompass_Preferred
Metaxalone-d3 is a deuterium labeled Metaxalone, an FDA-approved muscle relaxant. It primarily acts on the central nervous system to achieve muscle relaxation by inhibiting polysynaptic reflex arcs. This compound also inhibits MAO-A, demonstrating anti-inflammatory and antioxidant effects. It inhibits IL-1β-induced inflammatory phenotype, modulates NF-κB and other related signaling pathways, and decreases MAO-A expression and activity in IL-1β-treated microglia.
  • Deuterium labeled compound
  • FDA-approved muscle relaxant
  • Inhibits polysynaptic reflex arcs
  • Inhibits MAO-A
  • Provides anti-inflammatory and antioxidant effects
  • Modulates NF-κB and related signaling pathways
  • Decreases MAO-A expression and activity in IL-1β-treated microglia
  • Used as a tracer for quantitation in drug development
Catalog No. 50-004-09180