Learn More
Cayman Chemical Met-Enkephalinamide (trifluoroacetate salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502909199
Encompass_Preferred
A peptide opioid receptor agonist; binds to opioid receptors in rat cerebellum homogenates (IC50 = 0.2 µM); reduces the twitch height in electrically stimulated isolated guinea pig ileum and isolated mouse vas deferens; induces analgesia in the hot plate and tail-flick tests in rats at 300 µg/animal
Catalog No. 50-290-9199 Supplier Cayman Chemical Supplier No. 3608150 MG
A peptide opioid receptor agonist; binds to opioid receptors in rat cerebellum homogenates (IC50 = 0.2 µM); reduces the twitch height in electrically stimulated isolated guinea pig ileum and isolated mouse vas deferens; induces analgesia in the hot plate and tail-flick tests in rats at 300 µg/animal