Learn More
AdipoGen Meteorin-like (mouse) (rec.) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B01493010
Encompass_Preferred
Protein. Mouse meteorin-like (46-311) is fused at the N-terminus to a FLAG-tag. Source: HEK 293 cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). Meteorin-like (cometin; subfatin) is a potent new neurotrophic factor promoting neurite outgrowth from dorsal root ganglion cells. Meteorin-like is also a novel adipokine expressed by adipose tissue being downregulated upon caloric restriction. Meteorin-like is secreted by muscles during exercise and converts white adipose cells into brown fat tissue.
Catalog No. 50-205-3237