Learn More
Moldex™ Meteors™ Ecostation™ Ear Plug Refill
SDP
Catalog No. 19384375 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
33 dB
Catalog No. Noise Reduction Rating (NRR)
19-384-375 33 dB
1 options
Catalog No. 19-384-375 Supplier Moldex™ Supplier No. 6708
Add to Cart
Add to Cart
Convenient refill container for Meteors™ Ecostation™ dispenser (6707). Meteors™ unique curved design provides exceptional comfort. The contours follow the shape of the ear canal which means less pressure at the smallest diameter of the ear plug.
  • 100% PVC-free
  • Soft, slow recovery foam and contoured shape molds easily to ear canal for comfort
  • Contours follow ear canal shape for less pressure at the smallest diameter of the ear plug
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Regular
High Visibility Yes
Packaging Type Refill
Pairs per Package 500 Pairs/Dispenser Refill
Plug Material Foam
Plug Color Green
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No