Learn More
Cole-Parmer Myron L TPH1 TechPro II pH/Conductivity Meter
Catalog No. 50631809 Shop All Cole-Parmer Products
Encompass_Preferred
The TechPro II series of meters provide pH, conductivity, and TDS measurements for applications such as boilers and cooling towers, reverse osmosis, pools and spas, pulp and paper, and many other industries. The meters require only a 1.2-mL sample for pH measurement and a 5-mL sample for conductivity measurements. Both the temperature-compensated measurement of interest and the sample temperature is displayed on the four-digit LCD. The meters have autoranging function for greater resolution of your conductivity measurements between 0 and 9999 µS or 10 to 20.00 mS. The meter features an internal, non-refillable pH sensor that protects the glass pH bulb from breakage while allowing field replacement.
Catalog No. 50-631-809 Supplier Cole-Parmer Supplier No. 1970707
The TechPro II series of meters provide pH, conductivity, and TDS measurements for applications such as boilers and cooling towers, reverse osmosis, pools and spas, pulp and paper, and many other industries. The meters require only a 1.2-mL sample for pH measurement and a 5-mL sample for conductivity measurements. Both the temperature-compensated measurement of interest and the sample temperature is displayed on the four-digit LCD. The meters have autoranging function for greater resolution of your conductivity measurements between 0 and 9999 µS or 10 to 20.00 mS. The meter features an internal, non-refillable pH sensor that protects the glass pH bulb from breakage while allowing field replacement.