Learn More
Cole-Parmer Myron L EP-10 Analog Conductivity Meter 01489-35
Supplier:  Cole-Parmer 0148935
Encompass_Preferred
Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL.
Catalog No. 50-631-796
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov