Learn More
Cole-Parmer Myron L analog conductivity/resistivity meter
Catalog No. 50631795 Shop All Cole-Parmer Products
Encompass_Preferred
Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL.
Catalog No. 50-631-795 Supplier Cole-Parmer Supplier No. 0148930
Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL.