Learn More
Cole-Parmer Myron L Ultrameter 4PII Waterproof Conductivity/TDS/Resistivity Meter 19401-12
Catalog No. 50631773 Shop All Cole-Parmer Products
Encompass_Preferred
These meters are simple and easy to use just add your sample to the built-in cell cup, push the button, and take a reading. Autorange function automatically selects from five conductivity ranges. Factory-calibrated meters have conductivity-to-TDS conversion curves for NaCl, 442, and KCl standards stored in memory. You can also calibrate using your own calibration values. Automatic temperature compensation with adjustable temperature coefficients up to 9.99%/C provides accurate and precise measurements. You can easily store and recall up to 100 measurements with time-and-date stamp. Nonvolatile memory retains all data.
Catalog No. 50-631-773 Supplier Cole-Parmer Supplier No. 1940112
These meters are simple and easy to use just add your sample to the built-in cell cup, push the button, and take a reading. Autorange function automatically selects from five conductivity ranges. Factory-calibrated meters have conductivity-to-TDS conversion curves for NaCl, 442, and KCl standards stored in memory. You can also calibrate using your own calibration values. Automatic temperature compensation with adjustable temperature coefficients up to 9.99%/C provides accurate and precise measurements. You can easily store and recall up to 100 measurements with time-and-date stamp. Nonvolatile memory retains all data.