Learn More
Cole-Parmer Myron L 512T5 Analog Total Dissolved Solids Meter 01489-15
Catalog No. 50631775 Shop All Cole-Parmer Products
Encompass_Preferred
Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold below) expands range by a factor of 10. Minimum sample volume is 25 mL.
Catalog No. 50-631-775 Supplier Cole-Parmer Supplier No. 0148915
Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold below) expands range by a factor of 10. Minimum sample volume is 25 mL.