Learn More
Cole-Parmer Myron L 532T1 Analog Total Dissolved Solids Meter 01489-25
Catalog No. 50631777 Shop All Cole-Parmer Products
Encompass_Preferred
Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL.
Catalog No. 50-631-777 Supplier Cole-Parmer Supplier No. 0148925
Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL.