Learn More
Cole-Parmer Oakton Ion 6+ Handheld Meter Kit with NIST-Traceable Calibration
Catalog No. 501961348 Shop All Cole-Parmer Products
Encompass_Preferred
These rugged, economical meters are very easy to use. Features include all push-button calibration, hold function, auto-off, and splash-resistant keypad. Read concentration in ppm and mV as well as pH and temperature in C. The included protective rubber boot also acts as a meter stand for benchtop use. Order ISE electrodes separately.
Catalog No. 50-196-1348 Supplier Cole-Parmer Supplier No. 3561383
These rugged, economical meters are very easy to use. Features include all push-button calibration, hold function, auto-off, and splash-resistant keypad. Read concentration in ppm and mV as well as pH and temperature in C. The included protective rubber boot also acts as a meter stand for benchtop use. Order ISE electrodes separately.