Learn More
Cole-Parmer Extech 380460 Precision Four-Wire Milliohm Meter, 110 VAC, 60 Hz
Catalog No. 50993005 Shop All Cole-Parmer Products
Encompass
This precision milliohmeter features high accuracy and performance for low resistance measurements. The large 0.7" LCD shows 1999 count while still being built into compact, durable field service housing with protective cover.
Catalog No. 50-993-005 Supplier Cole-Parmer Supplier No. 2684900
This precision milliohmeter features high accuracy and performance for low resistance measurements. The large 0.7" LCD shows 1999 count while still being built into compact, durable field service housing with protective cover.