Learn More
Cole-Parmer Myron L EP11/PH Analog pH/Conductivity Meter
Catalog No. 50631797 Shop All Cole-Parmer Products
Encompass_Preferred
Get accurate, on-the-spot measurements of pH and conductivity with this compact meter. Meter features analog display of conductivity measurements from 0 to 10,000 µS in four ranges. Meter also includes a pH sensor that measures pH from 2.0 to 12.0 pH. Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL. The optional calibration kit 01489-51 includes a large foam-lined carrying case; 2-oz bottles of pH 4, 7 and 10 buffers; and a 7000 µS conductivity standard.
Catalog No. 50-631-797 Supplier Cole-Parmer Supplier No. 0148962
Get accurate, on-the-spot measurements of pH and conductivity with this compact meter. Meter features analog display of conductivity measurements from 0 to 10,000 µS in four ranges. Meter also includes a pH sensor that measures pH from 2.0 to 12.0 pH. Analyze your samples on-site with these portable meters. All feature automatic temperature compensation and built-in cell cup. Cup contains self-maintaining electrodes that never need replatinizing. Optional conductivity range extender (sold separately) expands range by a factor of 10. Minimum sample volume is 25 mL. The optional calibration kit 01489-51 includes a large foam-lined carrying case; 2-oz bottles of pH 4, 7 and 10 buffers; and a 7000 µS conductivity standard.