Learn More
Cole-Parmer Oakton pH 5+ Handheld Meter with NIST-Traceable Calibration
Catalog No. 501961345 Shop All Cole-Parmer Products
Encompass_Preferred
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80C. Meters include a protective rubber boot that also acts as a meter stand.
Catalog No. 50-196-1345 Supplier Cole-Parmer Supplier No. 3561351
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80C. Meters include a protective rubber boot that also acts as a meter stand.