Learn More
ACL Inc METER RESISTIVITY ACL 395
Catalog No. 501037856
Encompass_Preferred
Static control instruments, resistivity meters, ACL Staticide, model 395, Surface Resistivity Meter, LED scale measures 10^3 - 10^12 ohms, rubber rails, optional 5 lb probes available separately
This item is not returnable. View return policy
Catalog No. 50-103-7856 Supplier ACL Inc Supplier No. ACL 395
Static control instruments, resistivity meters, ACL Staticide, model 395, Surface Resistivity Meter, LED scale measures 10^3 - 10^12 ohms, rubber rails, optional 5 lb probes available separately