Learn More
Cole-Parmer Extech 407750 Integrating Sound Meter with RS-232
Supplier:  Cole-Parmer 4042500
Encompass
Meter features six sound level ranges, analog bar graph, and RS-232 interface for capturing data directly on your PC (order software separately). Use the selectable backlit display for easy readability in low kight conditions. What''s included: microphone windscreen, carrying case, and one 9 V battery. Compliance: ANSI S1.4-1983 Type 2, IEC 651 Type 2
Catalog No. 50-993-049