Learn More
Cole-Parmer Extech RH101 Thermohygrometer and IR Thermometer
Supplier:  Cole-Parmer 3780302
Encompass
Thermohygrometer simultaneously displays humidity and probe temperature, or humidity and infrared temperature. Measure temperature and humidity in laboratories, cleanrooms and storage areas with the remote probe. For high temperature applications or measurements in hard-to-reach areas, use the infrared thermometer. View measurements in low light areas with backlit display. Meter features switchable °F/°C units, separate MAX and HOLD buttons for temp/humidity and infrared measurements, low battery and overrange indicators, and auto shutoff. What’s included: protective holster, built-in stand, probe and one 9V battery.
Catalog No. 50-993-030