Learn More
Cole-Parmer Fluke 971 Thermohygrometer, 5 to 95% RH/, -4 to 140F
Supplier:  Cole-Parmer 3785120
Encompass
The Fluke 971 meter is built for field use with an impact resistant housing, rugged holster, convenient belt clip, and bright backlit display. Includes degF/degC selection, Min/Max reading, hold feature, averaging function, and 99 record datalogging capacity. A large LCD simultaneously displays RH and temperature readings, along with calculating dew point and wet bulb. Available for sale in the United States and Canada only.
Catalog No. 50-992-559