Learn More
Columbus Instruments Intl Corp Grip Strength Meter, Objective Neuromuscular Assessment, Digital Display, 1 Unit
SDP
Catalog No. NC3453221
Encompass
The Columbus Instruments Grip Strength Meter is a precision tool for assessing neuromuscular function and muscle strength in mice. It features a single-sensor design for objective measurement, with a digital display for instant force readouts. This meter includes advanced software for easy data management and CSV export, making it ideal for research in neurosciences and pharmaceutical studies.
Catalog No. NC3453221 Supplier Columbus Instruments Intl Corp Supplier No. 01678000
Add to Cart
Add to Cart
Request a Quote
The Columbus Instruments Grip Strength Meter is a precision tool for assessing neuromuscular function and muscle strength in mice. It features a single-sensor design for objective measurement, with a digital display for instant force readouts. This meter includes advanced software for easy data management and CSV export, making it ideal for research in neurosciences and pharmaceutical studies.