Learn More
Health Care Logistics Inc Metered Pump Nasal Spray Bottles, 1 oz. 12/pk
SDP
Catalog No. NC1389713
Encompass
Metered Pump Nasal Spray Bottles deliver 0.1cc for a consistent dose. Made of polyethylene.
This item is not returnable. View return policy
Catalog No. NC1389713 Supplier Health Care Logistics Inc Supplier No. 10271
Metered Pump Nasal Spray Bottles deliver 0.1cc for a consistent dose. Made of polyethylene.