Learn More
Med Vet International EasyTouch Glucose Meter, Each
Supplier:  Med Vet International 807002
Encompass_Preferred
Useful Features 100% Code Free 0.5 Microliter Blodd Sample Results in 5 seconds Altenate Site Testing 300 Most Recent Results 7/14/30 Day Averaging 4 Alarm Setting Contents EasyTouch Meter Instruction Manual Battery Included
Catalog No. 50-255-9671