Learn More
Cayman Chemical Metergoline, Purity: ≥98%, CAS: 17692-51-2, 10 mg
SDP
Catalog No. 502981370
Encompass_Preferred
A serotonin receptor antagonist active at 5-HT1 (pKi = 7.8 for 5-HT1B), 5-HT2, (pKi = 8.64, 8.75, and 8.75 for 5-HT2A, 5-HT2B, and 5-HT2C respectively), and 5-HT7 receptors; blocks 5-HT7 receptor-stimulated cAMP production (IC50 = 321 nM); dose-dependently inhibits voltage-dependent sodium and potassium channels
Catalog No. 50-298-1370 Supplier Cayman Chemical Supplier No. 2133610 MG
A serotonin receptor antagonist active at 5-HT1 (pKi = 7.8 for 5-HT1B), 5-HT2, (pKi = 8.64, 8.75, and 8.75 for 5-HT2A, 5-HT2B, and 5-HT2C respectively), and 5-HT7 receptors; blocks 5-HT7 receptor-stimulated cAMP production (IC50 = 321 nM); dose-dependently inhibits voltage-dependent sodium and potassium channels