Learn More
Thermo Scientific Capillary Metering Head, VHA1, 1 EA
Supplier:  Thermo Scientific 6850.1920
Encompass
A narrow bore capillary tube made of stainless steel or glass with inner diameter typically less than 1mm for precise fluid flow measurement and metering via pressure differential across the tube in applications including gas chromatography, mass flow controllers, leak detectors and microfluidic devices.
Catalog No. NC1319920