Learn More
Plasma Etch Inc METERING VALVE
SDP
Catalog No. NC2865646
Encompass
Metering Valve- recommended when processing lightweight parts to help preventthem from moving during vent
Catalog No. NC2865646 Supplier Plasma Etch Inc Supplier No. PE25MV
Metering Valve- recommended when processing lightweight parts to help preventthem from moving during vent