Learn More
TARGETMOL CHEMICALS INC Metformin 100MG
SDP
Catalog No. 502876487
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Metformin (1,1-Dimethylbiguanide) is an AMPK activator with blood-brain barrier permeability. Metformin may improve glycemic control by increasing insulin sensitivity and decreasing intestinal glucose uptake, and is commonly used in type 2 diabetes research. Purity 99.77%
Catalog No. 50-287-6487 Supplier TARGETMOL CHEMICALS INC Supplier No. T8526100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Metformin (1,1-Dimethylbiguanide) is an AMPK activator with blood-brain barrier permeability. Metformin may improve glycemic control by increasing insulin sensitivity and decreasing intestinal glucose uptake, and is commonly used in type 2 diabetes research. Purity 99.77%