Learn More
Medchemexpress LLC HY-B0627 25mg , Metformin CAS:657-24-9 Purity:>98%
SDP
Catalog No. 502258976
Encompass_Preferred
HY-B0627 25mg Metformin CAS: 657-24-9 Metformin (1,1-Dimethylbiguanide) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin can cross the blood-brain barrier and triggers autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-8976 Supplier Medchemexpress LLC Supplier No. HYB062725MG
HY-B0627 25mg Metformin CAS: 657-24-9 Metformin (1,1-Dimethylbiguanide) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin can cross the blood-brain barrier and triggers autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.