Learn More
Enzo Life Sciences Metformin (5g). CAS: 1115-70-4
SDP
Supplier:  Enzo Life Sciences ALX270432G005
Encompass_Preferred
Metformin is an antidiabetic agent. Reduces the cardiovascular complications of diabetes. Does not induce hypoglycemia. Inhibits hepatic gluconeogenesis via activation of the LKB1/AMPK pathway. Has antiproliferative effects in cancer cell lines. Alternative name: 1,1-Dimethylbiguanide . hydrochloride. Purity: ≥97% (Assay). Appearance: White crystalline powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8556