Learn More
Selleck Chemical LLC Metformin
SDP
Catalog No. 5000003388
Encompass_Preferred
Metformin (1 1-Dimethylbiguanide) a widely used drug for treatment of type 2 diabetes activates AMP-activated protein kinase (AMPK) in hepatocytes Metformin promotes mitophagy in mononuclear cells Metformin induces apoptosis of lung cancer cells through activating JNK/p38 MAPK pathway and GADD153
Catalog No. 50-000-03388 Supplier Selleck Chemical LLC Supplier No. S595810MM/1ML
Metformin (1 1-Dimethylbiguanide) a widely used drug for treatment of type 2 diabetes activates AMP-activated protein kinase (AMPK) in hepatocytes Metformin promotes mitophagy in mononuclear cells Metformin induces apoptosis of lung cancer cells through activating JNK/p38 MAPK pathway and GADD153