Learn More
Medchemexpress LLC Metformin-d6 hydrochloride | 1185166-01-1 | MFCD06658649 | 98.0% | 171.66 g/mol | C4H6D6ClN5 | 5 MG
SDP
Catalog No. 5000263357
Encompass_Preferred
Metformin-d6 hydrochloride is a deuterium-labeled metformin hydrochloride supplied as a research-grade analytical standard for use as an internal standard in quantitative LC-MS/GC-MS and tracer studies. For laboratory research use only; not for human consumption.
  • Deuterium-labeled analogue suitable as an internal standard for mass spectrometry.
  • High chemical purity: 98.0% as reported by supplier certificate of analysis.
  • High isotopic enrichment (99.8%), supporting accurate quantification and tracing.
  • Available as a solid in small analytical quantities for method development and validation.
  • Compatible with common analytical workflows including LC-MS and GC-MS.
Catalog No. 50-002-63357 Supplier Medchemexpress LLC Supplier No. HY1102285MG
Metformin-d6 hydrochloride is a deuterium-labeled metformin hydrochloride supplied as a research-grade analytical standard for use as an internal standard in quantitative LC-MS/GC-MS and tracer studies. For laboratory research use only; not for human consumption.
  • Deuterium-labeled analogue suitable as an internal standard for mass spectrometry.
  • High chemical purity: 98.0% as reported by supplier certificate of analysis.
  • High isotopic enrichment (99.8%), supporting accurate quantification and tracing.
  • Available as a solid in small analytical quantities for method development and validation.
  • Compatible with common analytical workflows including LC-MS and GC-MS.