Learn More
Selleck Chemical LLC Metformin HCl S1950-1g
SDP
Catalog No. 503109768
Encompass_Preferred
Metformin HCl (1 1-Dimethylbiguanide HCl) decreases hyperglycemia in hepatocytes primarily by suppressing glucose production by the liver (hepatic gluconeogenesis) Metformin promotes mitophagy in mononuclear cells Metformin induces apoptosis of lung cancer cells through activating JNK/p38 MAPK pathway and GADD153
Catalog No. 50-310-9768 Supplier Selleck Chemical LLC Supplier No. S19501G
Metformin HCl (1 1-Dimethylbiguanide HCl) decreases hyperglycemia in hepatocytes primarily by suppressing glucose production by the liver (hepatic gluconeogenesis) Metformin promotes mitophagy in mononuclear cells Metformin induces apoptosis of lung cancer cells through activating JNK/p38 MAPK pathway and GADD153