Learn More
Medchemexpress LLC HY-17471A 25mg , Metformin (hydrochloride) CAS:1115-70-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17471A25MG
Encompass_Preferred
HY-17471A 25mg Metformin (hydrochloride) CAS: 1115-70-4 Metformin hydrochloride (1,1-Dimethylbiguanide hydrochloride) inhibits the mitochondrial respiratory chain in the liver, leading to activation of AMPK, enhancing insulin sensitivity for type 2 diabetes research. Metformin hydrochloride triggers autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9554