Learn More
Medchemexpress LLC Metformin (hydrochloride) | 1115-70-4 | 100.0% | 165.62 | 100 MG
SDP
Catalog No. 5000386856
Encompass_Preferred
Metformin hydrochloride is a compound that inhibits the mitochondrial respiratory chain in the liver, leading to AMPK activation and enhanced insulin sensitivity, useful in type 2 diabetes research. It also lowers glucose by inhibiting Ras-related protein 1 (Rap1) in SF1 hypothalamic neurons and reduces liver oxidative stress, inflammation, and apoptosis. Additionally, it regulates autophagy and inhibits renal cell carcinoma growth by activating AMPK and inhibiting the mTOR pathway.
  • Inhibits mitochondrial respiratory chain
  • Activates AMPK and enhances insulin sensitivity
  • Used for type 2 diabetes research
  • Lowers glucose by inhibiting Rap1
  • Reduces liver oxidative stress, inflammation, and apoptosis
  • Regulates autophagy by activating AMPK
  • Inhibits mTOR signaling pathway
  • Induces tumor cell autophagy
  • Inhibits renal cell carcinoma growth in vitro and in vivo
Catalog No. 50-003-86856 Supplier Medchemexpress LLC Supplier No. HY17471A100MG
Metformin hydrochloride is a compound that inhibits the mitochondrial respiratory chain in the liver, leading to AMPK activation and enhanced insulin sensitivity, useful in type 2 diabetes research. It also lowers glucose by inhibiting Ras-related protein 1 (Rap1) in SF1 hypothalamic neurons and reduces liver oxidative stress, inflammation, and apoptosis. Additionally, it regulates autophagy and inhibits renal cell carcinoma growth by activating AMPK and inhibiting the mTOR pathway.
  • Inhibits mitochondrial respiratory chain
  • Activates AMPK and enhances insulin sensitivity
  • Used for type 2 diabetes research
  • Lowers glucose by inhibiting Rap1
  • Reduces liver oxidative stress, inflammation, and apoptosis
  • Regulates autophagy by activating AMPK
  • Inhibits mTOR signaling pathway
  • Induces tumor cell autophagy
  • Inhibits renal cell carcinoma growth in vitro and in vivo