Learn More
Medchemexpress LLC Metformin hydrochloride | 1115-70-4 | 99.9% | 165.62 | 25 MG
SDP
Catalog No. 5000383937
Encompass_Preferred
Metformin hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It inhibits the mitochondrial respiratory chain in the liver, leading to AMPK activation and enhancing insulin sensitivity, and can be used in the study of type 2 diabetes. It also exerts central glucose-lowering effects by inhibiting Ras-related protein 1 (Rap1) in SF1 hypothalamic neurons. Additionally, it inhibits liver oxidative stress, nitrosative stress, inflammation, and apoptosis caused by liver ischemia/reperfusion injury. It regulates the expression of autophagy-related proteins by activating AMPK and inhibiting the mTOR signaling pathway, thereby inducing tumor cell autophagy and inhibiting the growth of renal cell carcinoma in vitro and in vivo.
  • Analytical standard
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-83937 Supplier Medchemexpress LLC Supplier No. 25MGHY17471AR
Metformin hydrochloride (Standard) is an analytical standard intended for research and analytical applications. It inhibits the mitochondrial respiratory chain in the liver, leading to AMPK activation and enhancing insulin sensitivity, and can be used in the study of type 2 diabetes. It also exerts central glucose-lowering effects by inhibiting Ras-related protein 1 (Rap1) in SF1 hypothalamic neurons. Additionally, it inhibits liver oxidative stress, nitrosative stress, inflammation, and apoptosis caused by liver ischemia/reperfusion injury. It regulates the expression of autophagy-related proteins by activating AMPK and inhibiting the mTOR signaling pathway, thereby inducing tumor cell autophagy and inhibiting the growth of renal cell carcinoma in vitro and in vivo.
  • Analytical standard
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Appearance: Solid
  • Color: White to off-white