Learn More
Medchemexpress LLC Metformin hydrochloride | 1115-70-4 | MFCD00012582 | 100.0% | 165.62 | C4H12ClN5 | 10 MG
SDP
Catalog No. 5000298706
Encompass_Preferred
Metformin hydrochloride is the hydrochloride salt of metformin supplied as an analytical reference standard for use in analytical chemistry, method development, and research. It is identified by CAS 1115-70-4, has formula C4H12ClN5, and a molecular weight of 165.62.
  • High purity suitable for analytical assays (reported 99.97%).
  • Provided as a 10 MG package for calibration and method development.
  • Highly soluble in water and PBS (≥100 mg/mL); limited solubility in DMSO.
  • Storage: keep sealed at 4°C; follow solvent storage guidelines for long-term stability.
  • Intended for use as an analytical reference in validation, quality control, and research workflows.
Catalog No. 50-002-98706 Supplier Medchemexpress LLC Supplier No. HY17471AR10MG
Metformin hydrochloride is the hydrochloride salt of metformin supplied as an analytical reference standard for use in analytical chemistry, method development, and research. It is identified by CAS 1115-70-4, has formula C4H12ClN5, and a molecular weight of 165.62.
  • High purity suitable for analytical assays (reported 99.97%).
  • Provided as a 10 MG package for calibration and method development.
  • Highly soluble in water and PBS (≥100 mg/mL); limited solubility in DMSO.
  • Storage: keep sealed at 4°C; follow solvent storage guidelines for long-term stability.
  • Intended for use as an analytical reference in validation, quality control, and research workflows.