Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metformin Hydrochloride, BP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18604342 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-342 25 g Amber Glass Bottle
18-604-343 100 g Amber Glass Bottle
18-604-344 500 g Amber Glass Bottle
18-604-345 1 kg Amber Glass Bottle
18-604-346 5 kg Amber Glass Bottle
18-604-347 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-342 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M156625GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

M1566, 1115-70-4, C4H11N5·HCl

Spectrum™ Chemical Metformin Hydrochloride, BP is an antidiabetic used in the treatment of type 2 diabetes. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5
Molecular Weight (g/mol) 165.63
MDL Number MFCD00012582
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
IUPAC Name hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98.5 to 101%
Grade BP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Metformin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.