Learn More
Abcam Metformin hydrochloride (mM/ml), AMPK activator, 100MM_ML
Catalog No. 502704507
Encompass_Preferred
MW 165.63 g/mol, Purity >99%. AMPK activator. Antidiabetic and anticancer activity in vivo. Rapidly crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4507 Supplier Abcam Supplier No. AB146725100MM_ML
MW 165.63 g/mol, Purity >99%. AMPK activator. Antidiabetic and anticancer activity in vivo. Rapidly crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement