Learn More
U.S. Pharmacopeia Metformin Related Compound A, 461-58-5, MFCD00008066, 30mg
Catalog No. NC9261495
Encompass
Molecular formula C2H4N4, Molecular weight 84.1, Melting Point 409.1 - 411.8 °F (209.5 - 211 °C), Boiling Point 485.6 °F (252 °C) (solidified), Synonyms: Dicyanodiamide, 1-Cyanoguanidine
Catalog No. NC9261495 Supplier U.S. Pharmacopeia Supplier No. 1396310
Molecular formula C2H4N4, Molecular weight 84.1, Melting Point 409.1 - 411.8 °F (209.5 - 211 °C), Boiling Point 485.6 °F (252 °C) (solidified), Synonyms: Dicyanodiamide, 1-Cyanoguanidine