Learn More
Abcam Methylated DNA Immunoprecipitation (MeDIP) Kit - DNA, 48 Tests
Catalog No. NC2226212
Encompass_Preferred
Methylated DNA Immunoprecipitation (MeDIP) Kit - DNA (ab117133) enables the user to enrich methylated DNA by using an antibody specific to methyl cytosine (5-mC) to immunoprecipitate methylated genomic DNA (MeDIP).
The product is subject to the following: Abcam Restricted Use Statement
This item is not returnable. View return policy
Catalog No. NC2226212 Supplier Abcam Supplier No. AB11713348TEST
Methylated DNA Immunoprecipitation (MeDIP) Kit - DNA (ab117133) enables the user to enrich methylated DNA by using an antibody specific to methyl cytosine (5-mC) to immunoprecipitate methylated genomic DNA (MeDIP).
The product is subject to the following: Abcam Restricted Use Statement