Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methabenzthiazuron 98.0+%, TCI America™
SDP

Catalog No. M2334100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2334100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2334100MG Supplier TCI America Supplier No. M2334100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18691-97-9
Molecular Formula C10H11N3OS
Molecular Weight (g/mol) 221.28
MDL Number MFCD00055498
InChI Key RRVIAQKBTUQODI-UHFFFAOYSA-N
Synonym methabenzthiazuron, metabenzthiazuron, methbenzthiazuron, tribunil, preparation 5633, caswell no. 081b, 1,3-dimethyl-3-2-benzothiazolyl urea, bayer 5633, urea, n-2-benzothiazolyl-n,n'-dimethyl, unii-82npf43p0x
PubChem CID 29216
ChEBI CHEBI:6803
IUPAC Name 1-(1,3-benzothiazol-2-yl)-1,3-dimethylurea
SMILES CNC(=O)N(C)C1=NC2=CC=CC=C2S1

Specifications

Melting Point 121°C
Color White
Quantity 100 mg
UN Number 3077
Formula Weight 221.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methabenzthiazuron
RTECSNumber YR8980000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.